Would you like to log in again?
Sign, 'OPEN' Accuform®
Supplier: Accuform
MADM540VP MADM540VS MADM540VA
76441-710EA 28.18 USD InStock
76441-710 76441-712 76441-708
Sign, 'OPEN' Accuform®
Signs
Signs indicate when a facility is open and operational.
  • Multiple material options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty