Would you like to log in again?
Sign, 'DANGER KEEP OUT, CONTAMINATED AREA' Accuform®
Supplier: Accuform
MBHZ132VA MBHZ130VS MBHZ132VP MBHZ132VS MBHZ130VA MBHZ130VP
76441-742EA 17.15 USD InStock
76441-742 76441-740 76441-746 76441-744 76441-738 76441-748
Sign, 'DANGER KEEP OUT, CONTAMINATED AREA' Accuform®
Signs
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Multiple material and size options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty