Would you like to log in again?
Tags 'Biohazard Hazard Identity', Accuform
Supplier: Accuform
MDT926PTP MDT926CTP
76441-944PK 51.15 USD InStock
76441-944 76441-942
Tags 'Biohazard Hazard Identity', Accuform
Labels
Safety tags identify a temporary hazard until that hazard is eliminated or hazardous operation is complete.
  • Multiple material options available
Choose from two material options. PF-Cardstock tags are thin and flexible poly-based cardstock without wood content. They are durable, tear-resistant, waterproof, and will not crack or shrink, when compared to traditional paper cardstock. RP-Plastic tags are strong and durable plastic designed to resist chemicals, tears, and abrasions, plus withstand dirt and grease. The top of the RP-Plastic tags have a ³/₈" hole reinforced with a brass grommet to prevent pull-off (up to 50 lbs.).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty