Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Standard Clinical Centrifuge, Globe Scientific

Supplier: Globe Scientific
GCC-S
76462-044EA 567.11 USD InStock
76462-044
Standard Clinical Centrifuge, Globe Scientific
Centrifuges Benchtop Centrifuges
For simple, reliable operation, the GCC-S Standard Clinical Centrifuge simply can not be beat.

  • Run time: 1 sec - 99 min or continuous
  • Speed accuracy: ±100 rpm
  • Rotor capacity: 12×5/7/10 ml tubes or 8×15 ml tubes
  • Display: LED
  • Acceleration time: 23 s
  • Braking time: 23 s

The GCC-S Clinical Centrifuge has been designed for numerous clinical and research applications with simple yet robust features. The included 12-position rotor accommodates 5, 7 or 10 ml culture or blood collection tubes and up to 8×15 ml tubes (all adapters are included).

An easy to read LED display shows both time and speed and is easily programmed. The timer will not count down until the set speed is reached to ensure your separations and experiments work as intended. The manual door release is positioned on the side of the unit to ensure the GCC-S remains closed until you want access.

Certifications: CE, cTÜVus, FCC, MCA

Accessories information: Rotor risers and sleeves allow use of 5, 7 or 10 ml blood tubes.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty