Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cole-Parmer® Integrity 10 Reaction Station, Antylia Scientific

Supplier: Antylia Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

36630-00
75871-000EA 58540.81 USD InStock
75871-000
Cole-Parmer® Integrity 10 Reaction Station, Antylia Scientific
Stirrers Magnetic Hotplate Stirrers
Individually control temperature and stirring rate for each cell.

  • Perform 10 independent experiments at once saving time and money
  • View the results of your experiments in real time on the monitor
  • Easily transfer data to a PC via a USB port or SD card
  • Variable stirring speeds from 350 to 1200 rpm ensure homogeneous sample mixing
  • Set sample temperature between –30 and 150 °C with ±0.5 °C temperature accuracy

Use the Integrity 10 reaction station to run ten independent experiments at once. By setting the temperature and stir rate individually for each cell you can assess the impact of both at the same time, speeding up investigative chemistry by ten-fold.

Easily set up the test parameters using the user-friendly touchscreen menu. Intuitive icons reduce potential for costly errors while preprogrammed profiles for routine measurements saves time. Store results with information such as reaction composition, experiment owner, concentration, and the name of the profile with data stamp. View the progress of the reaction within each cell on the large monitor. It can also display at-a-glance results graphically. Extensive storage capacity lets you capture and review data. Download data to a PC via a USB port or SD card. Order software 36630-92 for more in-depth analysis and evaluation.

Control the temperature for each cell through the block or by using a temperature probe in each cell's solution. A ±0.5 °C temperature stability and a minimal set point overshoot of 0.5 °C ensure temperature accuracy. Set ramps between 0.1 °C/min to 5 °C/min for fast cooling and heating. Use the crash function for even faster temperature changes, which is ideal for kinetic determinations. Individually controlled magnetic stirrer can be set for speeds between 350 to 1200 rpm (use 12×4.5 mm cylindrical or 10×6 mm oval stir bars only). Order reducing sleeves and test tubes to accommodate sample volumes down to 1 ml.

The reflux/inerting head 36630-97 is ideal when you need to run parallel reactions for a long time, as is commonly required in organic chemistry experiments. Use with inerting caps to provide inert atmosphere to all ten cells. The integrated design eliminates the need to connect individual gas lines to each cell for a safer and effective solution.

Delivery information: Supplied with SD card, ten stir bars, and three power cords with US, UK, and European plugs.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty