Would you like to log in again?
Diversey™ Avert Sporicidal Disinfectant Cleaner Chlorine Wipes, Essendant
Supplier: Essendant
Diversey™
DVO100895790 DVO100895931
76461-978CT 585.57 USD InStock
76461-978 76461-976
Diversey™ Avert Sporicidal Disinfectant Cleaner Chlorine Wipes, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
Avert™ Sporicidal Disinfectant Cleaner Wipes clean, disinfect, and deodorize hard, non-porous surfaces in one labor-saving step with no rinsing required. Designed for use on hard, nonporous surfaces such as those found in hospitals and other healthcare facilities.
  • Effective in four minutes against Clostridium difficile (C. diff) spores
  • Heavy-duty disinfectant wipes clean, disinfect, and deodorize in one step
  • Bleach-based formula means no quat-binding concerns
  • No added fragrance or dyes
Avert Sporicidal Disinfectant Cleaner Chlorine Wipes are designed to clean and disinfect sinks, faucets, fixtures, bathrooms, restrooms, tubs, and showers.
Use these single-ply wipes on hard, non-porous surfaces to eliminate Clostridium Difficile dirt types.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty