Would you like to log in again?
Sargent Welch® Model Bundle- Plants
Supplier: Avantor
470154-968EA 375.09 USD InStock
470154-968
Sargent Welch® Model Bundle- Plants
Models Botany Models
Buy together and save.
  • Seven model set
  • Compare Monocot and Dicot structures
  • Hand painted
Easily compare Monocot and Dicot structures with this set of seven models. Each model is individually mounted on a backer, so they are easy to pass around. Included are stem root and leaf models for Monocot and Dicot structure. Also included is a generalized flower for floral anatomy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty