Would you like to log in again?
Tronex® AAMI Level 2 Chemotherapy Gowns
Supplier: Tronex
Tronex®
6333CHEMO35 6333CHEMO25
76462-946CS 130.31 USD InStock
76462-946 76462-944
Tronex® AAMI Level 2 Chemotherapy Gowns
Disposable Garments
Meeting the respective Association for the Advancement of Medical Instrumentation (AAMI) specifications for fluid resistance, this AAMI Level 2 chemo-rated gown features speacialized construction and high-comfort design and fit.
  • Soft, multi-layer, non-woven spunbond material with a fluid-impervious polyethylene coating for exceptional fluid and chemical resistance
  • Full front, long sleeves, waist ties, knitted cuffs, and a full back for complete coverage
  • Ultrasonically sealed seams for superior protection and high tensile strength
  • Not made with natural rubber latex
This quality chemo-rated gown provides AAMI Level 2 protection against fluid penetration, offering an ideal solution for procedures where exposure to blood, bodily fluids, chemicals, and other infectious substances might occur.
Certifications: Meets AAMI PB70 Level 2 Liquid Barrier Performance standards. Compliant with USP <800> Hazardous Drugs-Handling in Healthcare Settings. Chemo-rated material has passed permation testing with 12 chemotherapy drugs against ASTM standard F739.
Packaging: Gowns are packaged 10 per bag, 5 bags per case, for a total of 50 gowns.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty