Would you like to log in again?
Tyramide Amplification Buffer Plus, Biotium
Supplier: Biotium
22029 22029-T
76423-466EA 138.56 USD InStock
76423-466 76423-468
Tyramide Amplification Buffer Plus, Biotium
Protein Linking and Labeling Systems
An optimized buffer with enhanced sensitivity for tyramide signal amplification of immunostaining in cells or tissue sections.
  • For cell and tissue staining with tyramide signal amplification
  • Significantly improved brightness, specificity, and sensitivity
  • Compatible with CF® Dye and other tyramides
  • 250 or 1000 reaction sizes
  • 1× Tyramide amplification buffer plus
  • 30% Hydrogen peroxide
Tyramide amplification buffer plus is an optimized buffer with enhanced sensitivity for tyramide signal amplification of immunostaining in cells or tissue sections. It provides significantly improved brightness, specificity, and sensitivity compared to Biotium's original ready-to-use tyramide amplification buffer (76403-880, 76403-882).
Tyramide amplification buffer plus was developed as a new and improved version of Biotium's original ready-to-use tyramide amplification buffer (76403-880, 76403-882). It is also a component in all of Biotium's tyramide amplification kits.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty