Would you like to log in again?
HOBOnet Wireless Repeater
Supplier: ONSET COMPUTER CORPORATION SE
RXW-RPTR-900
470323-652EA 187.79 USD InStock
470323-652
HOBOnet Wireless Repeater
Data Loggers
The HOBOnet Repeater is designed to work with the HOBOnet Wireless Sensor Network. The repeater is ideal when there is an obstruction between wireless sensors and the HOBO RX3000 or the HOBO MicroRX station, or for extending the transmission range of wireless sensors in the network.
  • Repeater extends the HOBOnet wireless range by 450 to 600 meters
  • Simple push button setup for joining a HOBOnet wireless network
  • Powered by a rechargeable AA batteries and built-In solar panel or lithium batteries
  • Operates in an outdoor environment with a solar radiation shield
Ordering information: A complete system requires at least one HOBO RX3000 Remote Monitoring Station, a HOBOnet Wireless Manager, and a HOBOnet Wireless Sensor, OR one HOBO MicroRX Station (which has an integrated HOBOnet Wireless Manager) and a HOBOnet Wireless Sensor. HOBOnet Wireless Repeaters can be added to extend the range.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty