Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol 8,000, flakes

Supplier: Electron Microscopy Sciences
Synonyms: PEG 8,000

19770
100504-218EA 154.26 USD InStock
100504-218
Polyethylene glycol 8,000, flakes
Polyethylene glycol 8,000
An embedding medium for microscopy and histochemistry


Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Density: 1.0845 (60 °C)
MDL Number: MFCD01779615
CAS Number: 25322-68-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty