Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide, powder 15.0 µm

Supplier: Electron Microscopy Sciences
Synonyms: Aluminium(III) oxideAlumina C

50362-15
100491-282EA 33.18 USD InStock
100491-282
Aluminium oxide, powder 15.0 µm
Aluminium oxide
These are abrasive powders that are prepared from aluminum oxide, offering excellent coarse and intermediate polishing, with fast results.


Formula: Al₂O₃
MW: 101.96 g/mol
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty