Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide, α-phase Type DX, powder 0.3 µm

Supplier: Electron Microscopy Sciences
Synonyms: ɑ-Tonerdeα-Alumina

50361-05
100501-322LB 120.65 USD InStock
100501-322
Aluminium oxide, α-phase Type DX, powder 0.3 µm
Aluminium oxide, α-phase
EMS's Alumina Powders are accurately controlled for all specs and particle size distribution, assuring you the best polishing results.

The following micron sizes are available: 0.05, 0.3, and 1.0.

Type DX Alumina Powder is specially treated to reduce the number of agglomerates (group of particles in each grade of manufactured alumina). The result is fast polishing and improved quality of the finished products.
Type N Alumina Powder is recommended when these improvements are not necessary.

Typical applications are:
Precision Optics
Metallographic Sections
Electro-Optical Crystals
Silicone Wafers
Ferrite Components
Gem Stones
Acrylic lenses


Formula: Al₂O₃
MW: 101.96 g/mol
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty