Would you like to log in again?
Aldehyde Dehydrogenase 2 Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB283364-100TEST
76452-690EA 763.01 USD InStock
76452-690
Aldehyde Dehydrogenase 2 Inhibitor Screening Kit (Fluorometric), BioVision
Assays Immunological Assays
For screening, studying and characterizing potential inhibitors of ALDH2.
  • A quick and sensitive way of screening, studying and characterizing potential inhibitors of ALDH2
Aldehyde Dehydrogenase 2 (ALDH2), mitochondrial is a member of the ALDH superfamily that participates in multiple metabolic pathways including the oxidation of toxic biogenic and environmental aldehydes. ALDH2 is a key enzyme located in the mitochondrial matrix. It plays an important role in ethanol metabolism by converting acetaldehyde, a carcinogenic metabolite of ethanol to acetate. Imbalances of ALDH2 have been linked to both alcoholism and alcohol sensitivity in people. Reduced ALDH2 activity has been strongly correlated with reduced alcohol consumption and reduced incidences of alcoholism, which led to the development of selective ALDH2 inhibitors as antidipsotropic or alcohol aversive agents for treating alcoholism. Disulfiram, an alcohol-aversive drug inhibiting ALDH2 has been approved by the FDA and is used clinically for over 40 years for treating alcohol dependency.
However, the drug is a non-specific alkylating agent leading to many adverse side effects and drug-drug interactions. Thus, novel and specific inhibitors of ALDH2 free of the negative side effects could be used in aversion therapy. BioVision’s Aldehyde Dehydrogenase 2 Inhibitor Screening Kit provides a quick and sensitive way of screening, studying and characterizing potential inhibitors of ALDH2. In this assay, acetaldehyde is oxidized by ALDH2 to form NADH, which then couples with PicoProbe™ to generate a fluorescence signal measured at Ex/Em = 535/587 nm. The percentage inhibition can be determined by comparing the activity of ALDH2 treated with test inhibitor(s) vs untreated control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty