Would you like to log in again?
Hydroxyurea Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB282919-100TEST
76452-694EA 809.86 USD InStock
76452-694
Hydroxyurea Colorimetric Assay Kit, BioVision
Assays Cellular Assays
To measure HU concentrations in human serum or plasma.
  • It can detect as low as 2 nmole of HU under the assay conditions
  • Is the first, commerically available kit to measure HU concentrations in human serum or plasma
  • Simple, rapid, sensitive and convenient way to measure HU concentrations in samples
Hydroxyurea (HU), also known as carbamohydroxamic acid is a small molecule drug and is used in a number of myeloproliferative, neoplastic, and non-hematological diseases. HU can promote hemoglobin synthesis, reduce thrombocytosis and is an anti-cancer agent for treating chronic myeloid leukemia, melanoma, head and neck cancer and refractory ovarian cancer. HU is also approved by FDA for treating Sickle Cell Disease. Additionally, HU is a ribonucleotide reductase inhibitor and blocks DNA replication and cell division. HU has some side effects including nausea, vomiting, diarrhea, sore throat, ulcers, fever, cough and shortness of breath. Thus, it is important to quantify HU concentration in serum or plasma. BioVision’s Hydroxyurea Colorimetric Assay Kit is the first, commerically available kit to measure HU concentrations in human serum or plasma. In this assay, HU is reduced by a series of redox reactions to form an intermediate, which in the presence of detection reagents generates a colored product measured at 540 nm. The colored signal is directly proportional to the HU concentration in the sample. The kit offers a simple, rapid, sensitive and convenient way to measure HU concentrations in samples. It can detect as low as 2 nmole of HU under the assay conditions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty