Would you like to log in again?
Human GAPDH Activity Assay Kit II, BioVision
Supplier: BioVision
AB284542-100TEST
76452-696EA 461.82 USD InStock
76452-696
Human GAPDH Activity Assay Kit II, BioVision
Assays Cellular Assays
For monitoring GAPDH activity.
  • The kit can measure as low as 20 μU ST activity under assay conditions,A simple, two-step plate based assay that measures the fluorescent product at Ex/Em = 410/470 nm
  • For measuring Sialyltransferase activity using a 96-well plate format,The kit can detect activity from both α-1,6 and α-2,6 sialyltransferases
Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH; EC 1.2.1.12) catalyzes the conversion of Glyceraldehyde-3-Phosphate (GAP) to 1, 3-Bisphosphate Glycerate (BPG) and plays a key role in glycolysis. The enzyme is involved in cellular processes such as apoptosis, membrane trafficking, iron metabolism and nuclear translocation. GAPDH is a stable metabolic enzyme and is constitutively expressed. Therefore, it is considered as a housekeeping gene. Deregulation of GAPDH activity is associated with cell proliferation and carcinogenesis. Accurate detection of GAPDH activity is important for diagnosing diseases and studying normal cellular physiology. BioVision’s Human GAPDH Activity Assay Kit II provides a simple and sensitive method for monitoring GAPDH activity in various sample types. In this assay, GAPDH catalyzes GAP into BPG and an intermediate, which then reacts with the developer to form a colored product that is measured at 450 nm. Our high-throughput adaptable assay can detect GAPDH activity as low as 10 mU/mL in a variety of samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty