Would you like to log in again?
SARS-CoV-2 S1 Protein-ACE2 Binding Inhibitor Screening Kit, BioVision
Supplier: BioVision
AB283370-100TEST
76452-702EA 1217.07 USD InStock
76452-702
SARS-CoV-2 S1 Protein-ACE2 Binding Inhibitor Screening Kit, BioVision
Assays Protein Assays
To screen for potential inhibitors of S1 protein binding to human ACE2.
  • Simple and easy to use
  • Provides a reliable test for high throughput screening of potential inhibitors of S1 protein binding to Human ACE2
  • The assay kit is adapted to a 96-well format
Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2), also known as the 2019 Novel Coronavirus (2019-nCoV) or human coronavirus 2019 (HCoV-19 or hCoV-19), is the cause of the Coronavirus Disease 2019 (COVID-19) pandemic. It is a RNA virus that causes severe respiratory diseases in humans. SARS-CoV-2 coronavirus contains four main structural proteins namely Spike (S), Membrane (M), Envelope (E), and Nucleocapsid (N) protein. Spike protein is located on the outer envelope of the virion and mediates the viral entry and thus, plays an important role in inducing neutralizing antibodies and protective immunity. S protein consists of S1 and S2 subunits. The S1 subunit contains a receptor-binding domain that can specifically bind to the host-receptor namely Angiotensin Converting Enzyme 2 (ACE2), which facilitates the entry of the virus into the target cells including respiratory or intestinal epithelial cells, endothelial cells, alveolar monocytes or macrophages.
The receptor recognition step is an important determinant of the viral infectivity, pathogenesis and host range. Therefore, an intervention strategy that targets S1 protein and ACE2 interaction presents an important target for vaccination or antiviral strategies that includes small molecules and therapeutic antibodies. BioVision’s SARS-CoV-2 S1 Protein-ACE2 Binding Inhibitor Screening Kit can be used to screen for potential inhibitors of S1 protein binding to human ACE2. In this assay, the binding of S1 protein to biotinylated human ACE2 is detected using Streptavidin-HRP. Subsequently, a TMB substrate is added to visualize the HRP enzymatic reaction thereby generating a blue colored product that changes to yellow once the stop solution is added. The density of the yellow color is proportional to the binding of S1 protein to Human ACE2. However, in the presence of potent inhibitor(s), the binding of S1 protein to Human ACE2 is suppressed thereby preventing the color generation. The assay kit is adapted to a 96-well format and provides a reliable test for high throughput screening of potential inhibitors of S1 protein binding to Human ACE2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty