Would you like to log in again?
Forensic Investigation: Stages of Decomposition in Fetal Pigs
Supplier: CYRGUS CO INC SE
PK-69FI
470332-248KT 46.3 USD InStock
470332-248
Forensic Investigation: Stages of Decomposition in Fetal Pigs
Specimens Preserved Specimens Preserved Animal Materials
In this activity, students will observe and study the process of decomposition using a fresh fetal pig.
  • Includes fresh pig and materials
  • Guide to the five stages of decomposition
  • Complete study of decomposition in under 4 weeks
Although admittedly not for the faint of heart, this activity brings the world of forensic science to life in an unfgorgettable way. Experience the process of decomposition first-hand with a real decomposing fetal pig. Requires an undisturbed outdoor area to lay out the fresh, fetal pig.
Number of setups: 1
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty