Would you like to log in again?
Mouse BDNF ELISA Kit EZ-Set (DIY Anti-body Pairs), Boster
Supplier: Boster
EZ0309
76467-114EA 252.09 USD InStock
76467-114
Mouse BDNF ELISA Kit EZ-Set (DIY Anti-body Pairs), Boster
Assays ELISAs
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects BDNF in Mouse samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty