Would you like to log in again?
Clorox® Total 360® Disinfectant Cleaner, Essendant
Supplier: Essendant
CLO60025EA CLO31650
76468-934CT 197.73 USD InStock
76468-934 76468-932
Clorox® Total 360® Disinfectant Cleaner, Essendant
Cleaning Agents Disinfectants
CloroxPro™ Total 360® Disinfectant Cleaner is designed to be paired with the Clorox® Total 360® Electrostatic Sprayer to clean, disinfect and deodorize. This disinfectant cleaner kills 99.9% of bacteria in five seconds and is EPA registered to kill 44 organisms including cold and flu viruses (Rhinovirus, Influenza A), Methicillin-resistant Staphylococcus aureus (MRSA), Norovirus, Salmonella enterica, Pseudomonas aeruginosa, Staphylococcus aureus (Staph) and Vancomycin-resistant Enterococcus faecium (VRE) in two minutes.
  • Clean, sanitize, kill 99.9% of bacteria in five seconds, and deodorize hard and soft surfaces in one simple step.
  • Designed to sanitize soft surfaces.
  • Use on a variety of hard, nonporous surfaces such as porcelain, stainless steel, tile, plastic, and other non-food contact surfaces
  • Inhibits the growth of mold and mildew for up to seven days
  • Eliminates odors
Safe to use on a variety of hard non-porous surfaces such as porcelain, stainless steel, tile, plastic, and other nonfood contact surfaces (when use as directed). It eliminates odors, inhibit the growth of mold and mildew for up to seven days and sanitizes soft surfaces. CloroxPro™ Total 360® Disinfectant Cleaner is a bactericidal in the presence of organic soil (5% blood serum). Approved for use with the Clorox ® Total 360® Electrostatic Sprayer and intended for use with schools, athletic facilities, offices, healthcare facilities, public restrooms, hotels, airports, ambulances, daycare centers, and institutional kitchens. CloroxPro™, where clean means everything. Packaging may vary.
Application: All Purpose; Applicable Material: Ceramics; Enamel; Fiberglass; Finished/Sealed Wood; Laminates; Metal; Plastic; Sealed/Engineered Stone; Vinyl.
Chemical Compound: Alkyl (60% C14, 30% C16, 5% C12, 5% C18) Dimethyl Benzyl Ammonium Chlorides 0.15%; Alkyl (68% C12, 32% C14) Dimethyl Ethybenzyl Ammonium Chlorides 0.15%.
Dirt Types: Bacteria; Cold and Flu Viruses; Dust; Fungus; Germs; Grease; Grime; MRSA; Mildew; Mold; Rhinovirus; Viruses; Water-Based Soil.
Accessories information: The CloroxPro™ Total 360® Electrostatic Sprayer, with the patented PowerWrap™ nozzle, delivers superior surface coverage for healthier facilities. The electrostatic technology charges, atomizes and delivers consistent wraparound coverage of solution to all surfaces – front, back and sides. Even those hard-to-reach, difficult-to-clean areas can now be properly disinfected and sanitized. Ideal for use in schools, athletic facilities, offices, healthcare facilities, public restrooms, hotels, airports, ambulances, daycare centers, institutional kitchens, correctional facilities - anywhere illness causing germs can quickly spread. Approved for use with CloroxPro™ Total 360® Disinfectant Cleaner, CloroxPro™ Anywhere® Hard Surface Sanitizing Spray or Clorox Healthcare® Spore Defense Spray. Ideal for use in schools, athletic facilities, offices, healthcare facilities, public restrooms, hotels, airports, ambulances, daycare centers, and institutional kitchens. CloroxPro™, where clean means everything.
Ordering information: Cart System-Ergonomically designed with lightweight handle. 12.5" x 19" x 32". Sprayer/Hose/Nozzle Type: Sprayer; Sprayer/Nozzle Type: Trigger; Nozzle/Sprayer Settings: Stream; Hose Diameter: 1".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty