Would you like to log in again?
EcoGII Methyltransferase
Supplier: New England Biolabs
M0603S
103391-812EA 84 USD InStock
103391-812
EcoGII Methyltransferase
Enzymes
EcoGII Methyltransferase is a non-specific methyltransferase that modifies adenine residues (N6) in any sequence context.
  • Non-specific methyltransferase that modifies adenine residues (N6) in any sequence context
  • Enzyme for Innovation (EFI) - part of a project initiated by NEB to provide unique enzymes
Delivery information: Supplied with 10X CutSmart buffer and 32 mM S-adenosylmethionine (SAM).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty