Would you like to log in again?
5-hydroxythymidine DNA Kinase, New England Biolabs
Supplier: New England Biolabs
M0659S M0659S
76284-684EA 81 USD InStock
76284-684 CA76284-684
5-hydroxythymidine DNA Kinase, New England Biolabs
Enzymes
5-hydroxymethyluridine DNA Kinase (5-HMUDK) transfers the gamma phosphate from ATP to the hydroxymethyl moiety of 5-hydroxymethyluridine in polymeric DNA.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty