Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD209 Rabbit Polyclonal Antibody (DyL550)

Supplier: Boster
A01025-DYL550
76464-098EA 378.14 USD InStock
76464-098
Anti-CD209 Rabbit Polyclonal Antibody (DyL550)
Antibodies
Rabbit IgG Polyclonal Anti-Human DC-SIGN Antibody DyLight®, 550 Conjugated, Flow Validated.

Type: Primary
Antigen: CD209
Clonality: Polyclonal
Clone:
Conjugation: DyL550
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty