Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-tfpia Rabbit Polyclonal Antibody

Supplier: Boster
DZ33957
76466-282EA 401.06 USD InStock
76466-282
Anti-tfpia Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for tfpia detection.

Type: Primary
Antigen: tfpia
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Zebrafish

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty