Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TCP1 Mouse Monoclonal Antibody [clone: 2] (DyL550)

Supplier: Boster
M02389-DYL550
76467-850EA 378.14 USD InStock
76467-850
Anti-TCP1 Mouse Monoclonal Antibody [clone: 2] (DyL550)
Antibodies
Mouse IgG Monoclonal Anti-Human TCP1 alpha Antibody DyLight®, 550 Conjugated, Flow Validated.

Type: Primary
Antigen: TCP1
Clonality: Monoclonal
Clone: 20000000
Conjugation: DyL550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty