Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYCS Mouse Monoclonal Antibody [clone: 15F10] (DyL488)

Supplier: Boster
M03529-DYL488
76467-906EA 378.14 USD InStock
76467-906
Anti-CYCS Mouse Monoclonal Antibody [clone: 15F10] (DyL488)
Antibodies
Mouse IgG Monoclonal Anti-Human Cytochrome C Antibody DyLight®, 488 Conjugated, Flow Validated.

Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: 15F10
Conjugation: DyL488
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty