Would you like to log in again?
Choline Oxidase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB284569-100TEST
76469-550EA 977.18 USD InStock
76469-550
Choline Oxidase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
To determine the activity of choline oxidase in samples.
  • The assay is simple, fast, reproducible and is high throughput adaptable
  • The first commercially available kit to determine the activity of choline oxidase in samples
  • The kit can detect as low as 0.2 mU of choline oxidase activity under assay conditions
Choline Oxidase (E.C. 1.1.3.17) belongs to the oxidoreductase family and is present in plants, bacteria, fungi and mammals. It is a catabolic enzyme that catalyzes the chemical reaction of choline and oxygen into betaine glycine and hydrogen peroxide with betaine aldehyde as an intermediate. The enzyme participates in glycine, serine, and threonine metabolism and is commonly used as a tool for the enzymatic determination of acetylcholinesterase, phospholipase D, phosphatidylcholine-specific phospholipase C, sphingomyelinase etc. Betaine glycine is a well known osmoprotectant and can help plants and bacteria to survive dry, cold and salt stresses. As a result, choline oxidase has been engineered genetically to produce transgenic plants such as tomato, rice and tobacco with increased stress tolerance and productivity.
Additionally, accumulation of betaine glycine has been observed in bacteria such as Staphylocccus aureus, Pseudomonas aeruginosa and Listeria monocytogenes thereby suggesting that drugs capable of inhibiting choline oxidase activity will have potential anti-bacterial activity. BioVison’s Choline Oxidase Activity Assay Kit is the first, commercially available kit to determine the activity of choline oxidase in samples. In this assay, choline oxidase oxidizes choline to generate hydrogen peroxide, which then reacts with the probe to generate a strong fluorescent signal measured at Ex/Em = 535/587 nm. The fluorescent signal is directly proportional to the choline oxidase activity in the samples. The assay is simple, fast, reproducible and is high throughput adaptable. The kit can detect as low as 0.2 mU of choline oxidase activity under assay conditions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty