Would you like to log in again?
Mycoplasma Staining Kit, BioVision
Supplier: BioVision
AB284574-10TEST AB284574-2TEST
76469-544EA 421.66 USD InStock
76469-544 76469-546
Mycoplasma Staining Kit, BioVision
Stains Microbiology Stains
A fluorescence based microscopic assay.
  • Ready-to-use
  • Ultrasensitive
  • Rapid results in 60 min or less
  • Visual identification
BioVision's Mycoplasma Staining Kit provides an ultrasensitive, rapid and simple fluorescence based microscopic assay for the visual identification of mycoplasma infections in laboratory cell cultures. Mycoplasma infections are relatively common in laboratory cell cultures. It has been estimated that between 5% and 35% of all cell cultures are infected. Mycoplasmas have been shown to alter the growth rate of cells in culture, induce chromosomal aberrations, influence amino acid and nucleic acid metabolism and cause membrane aberrations.
BioVision’s Mycoplasma Staining Kit contains a MycoFluor Reagent and Antifade Mounting Medium. The MycoFluor reagent is added directly to the culture medium, with or without cells, and the stained sample is then examined under a fluorescence microscope with an excitation and emission maximam of 350 to 360 nm and 450 to 460 nm respectively. Mycoplasma staining with MycoFluor reagent appears as fine particulate or filamentous staining in the cytoplasm at 100X magnification. Nuclei of the cells are also brightly stained by this method and thereby act as endogenous positive control for the staining procedure.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty