Would you like to log in again?
Human PDGF-AB PicoKine; Quick ELISA Kit, Boster
Supplier: Boster
PicoKine
FEK0484
76467-346EA 571.78 USD InStock
76467-346
Human PDGF-AB PicoKine; Quick ELISA Kit, Boster
Assays ELISAs
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Human Pdgf Bb/PDGFB with <2 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Pdgf Bb/PDGFB tissue specificity: Expressed at high levels in the heart, brain (sustantia nigra), placenta and fetal kidney. Expressed at moderate levels in the brain (hippocampus), skeletal muscle, kidney and lung.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty