Would you like to log in again?
Human Angiopoietin-1 / ANG1 PicoKine; Quick ELISA Kit, Boster
Supplier: Boster
PicoKine
FEK0559
76467-366EA 571.78 USD InStock
76467-366
Human Angiopoietin-1 / ANG1 PicoKine; Quick ELISA Kit, Boster
Assays ELISAs
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Human Angiopoietin-1 / ANG1 with <12 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Angiopoietin-1 / ANG1 tissue specificity: Expressed predominantly in the liver. Also The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detected in endothelial cells and spinal cord neurons.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty