Would you like to log in again?
Mouse Gp130/IL6ST PicoKine; Quick ELISA Kit, Boster
Supplier: Boster
PicoKine
FEK0577
76467-370EA 571.78 USD InStock
76467-370
Mouse Gp130/IL6ST PicoKine; Quick ELISA Kit, Boster
Assays ELISAs
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Mouse Gp130/IL6ST with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum and plasma (heparin, EDTA, citrate). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Gp130/IL6ST tissue specificity: Found in tissues such as brain, heart, thymus, spleen, kidney, lung and liver. Found in all the cell lines tested except BaF-B03. Expression not restricted to IL6-responsive cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty