Would you like to log in again?
Human Cystatin C/CST3 PicoKine; Quick ELISA Kit, Boster
Supplier: Boster
PicoKine
FEK0678
76467-382EA 571.78 USD InStock
76467-382
Human Cystatin C/CST3 PicoKine; Quick ELISA Kit, Boster
Assays ELISAs
Detect Human Cystatin C/CST3 with <10 pg/ml sensitivity.
Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum, plasma (heparin, EDTA), saliva, urine and human milk. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Cystatin C/CST3 tissue specificity: Expressed in submandibular and sublingual saliva but not in parotid saliva (at protein level). Expressed in various body fluids, such as the cerebrospinal fluid and plasma. Expressed in highest levels in the epididymis, vas deferens, brain, thymus, and ovary and the lowest in the submandibular gland.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty