Would you like to log in again?
Signs, 'CONTAINS VACCINE, DO NOT UNPLUG', Accuform®
Supplier: Accuform
MBDX509VS MBDX510VA MBDX509VA MBDX510VP MBDX509VP MBDX510VS
76471-092EA 9.25 USD InStock
76471-092 76471-094 76471-096 76471-098 76471-100 76471-102
Signs, 'CONTAINS VACCINE, DO NOT UNPLUG', Accuform®
Signs
Vaccination signs communicate health and safety policies, reinforce procedures, and identify locations.
  • Multiple material and size options available
Choose from three material options. Adhesive vinyl signs feature a printed matte surface with permanent acrylic adhesive back. These are moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Plastic is a good material choice for indoor and sheltered outdoor environments, as the printed surface has clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. They feature a printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions. They also offer abrasion and chemical resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty