Would you like to log in again?
Cy3 Conjugated Goat Anti-Mouse IgG SABC Kit, Boster
Supplier: Boster
SA1072
76468-058EA 343.76 USD InStock
76468-058
Cy3 Conjugated Goat Anti-Mouse IgG SABC Kit, Boster
Assays Immunological Assays
Cy3 Conjugated Anti-Mouse IgG SABC Kit is a four-component system for indirect sensitive immunofluorescent detection of proteins on tissue sections via ICC, IHC-P or IHC-F probed with mouse primary antibody by utilizing a biotinylated secondary antibody that links target-bound primary antibodies to streptavidin-Cy3 conjugate forming a stable StreptAvidin-Biotin Complex (SABC).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty