Would you like to log in again?
HRP Conjugated Rabbit Anti-Goat IgG SABC Kit, Boster Biological Technology
Supplier: Boster
SA2003
76468-098EA 320.84 USD InStock
76468-098
HRP Conjugated Rabbit Anti-Goat IgG SABC Kit, Boster Biological Technology
Assays Immunological Assays
HRP Conjugated anti-Goat IgG SABC Kit, with the specific secondary antibody and all the reagents needed for performing IHC-P;IHC-F;ICC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty