Would you like to log in again?
Tte-UvrD Helicase
Supplier: New England Biolabs
M1202S
76326-038EA 81 USD InStock
76326-038
Tte-UvrD Helicase
Enzymes
Tte UvrD Helicase is a repair helicase capable of unwinding double-stranded DNA, without a requirement for a specific flap or overhang structure, from the thermophilic organism Thermoanaerobacter tengcongensis.
  • Unwinds double-stranded DNA
  • Thermostable to 65 °C
  • Reduces non-specific product formation in isothermal amplification (LAMP)
It is active on a wide range of DNA substrates and, along with its thermostability (active to 70 °C), Tte UvrD Helicase has been demonstrated to be a useful additive for improving specificity of isothermal amplification reactions, particularly in conjunction with the WarmStart LAMP kit.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty