Would you like to log in again?
NEBExpress GamS Nuclease Inhibitor
Supplier: New England Biolabs
P0774S P0774S
76407-366EA 102 USD InStock
CA76407-366 76407-366
NEBExpress GamS Nuclease Inhibitor
Proteins and Peptides
NEBExpress GamS nuclease inhibitor is a recombinant protein that inhibits Exonuclease V (RecBCD) activity and stabilizes linear DNA templates in E. coli based in vitro protein synthesis reactions.
  • Enhances synthesis yield of linear DNA using NEBExpress Cell-free E. coli protein synthesis system
  • Recombinant enzyme; ≥95% purity, as determined by SDS-PAGE
  • No detectable protease activity, ensuring stability of desired target protein
  • Compatible with various target proteins, ranging from 17 to 230 kDa
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty