Would you like to log in again?
NEBExpress Ni Resin, New England Biolabs
Supplier: New England Biolabs
S1428S
76377-486EA 358 USD InStock
76377-486
NEBExpress Ni Resin, New England Biolabs
Filters Centrifugal Filters
NEBExpress Ni Resin is an affinity matrix for the isolation and purification of polyhistidine-tagged (His-tagged) fusion proteins.
  • Purification of >10 mg His-tagged protein per 1 ml of resinIntended for use in gravity or pressure flow columns, and batch purifications
  • High specific binding of His-tagged proteins yielding purities of >95%
  • Strong nickel ion binding provides excellent resistance to EDTA and reducing agents
It is intended for use in gravity or pressure flow columns, and batch purifications. NEBExpress Ni Resin is comprised of a highly uniform and stable chemical-tolerant resin, pre-charged with nickel ions on the matrix surface. It is resistant to a wide range of chemicals, including NaOH, EDTA, DTT and beta-Mercaptoethanol.
Concentration: 50%
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty