Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Drop-n-Stain EverBrite™ Mounting Medium, Biotium
Supplier: Biotium
23009 23008-T 23008 23009-T
76403-888EA 201.27 USD InStock
76403-888 76403-886 76403-890 76403-884
Drop-n-Stain EverBrite™ Mounting Medium, Biotium
Stains Microscopy and staining reagents
Wet-set antifade mounting medium for preserving fluorescence of our CF® dyes and other fluorochromes, in a dropper bottle for easy dispensing.
  • Convenient dropper bottle for pipette-free mounting
  • Protects fluorescent dyes from rapid photobleaching
  • Compatible with cyanine-based fluorophores like Cy®3, Cy®5, and Alexa Fluor® 647, unlike VECTASHIELD®
  • Available with or without DAPI for nuclear counterstaining
  • Works as well as original EverBrite™, but less viscous for rapid DAPI staining
Drop-n-Stain EverBrite™ mounting medium is a wet-set antifade mounting medium optimally formulated for preserving fluorescence of our CF® dyes and other fluorochromes. The dropper bottle makes for quick and easy dispensing.
It is a wet-set medium that remains liquid. After mounting, coverslips can be sealed with nail polish or CoverGrip™ coverslip sealant (89411-108). Mounted slides can be stored at 4 °C or –20 °C, protected from light. Antibody-stained slides can be preserved for a year or longer (note: some labels, like phalloidin, are less stable during storage). It has a refractive index of 1.42.
They also offer EverBrite™ hardset mounting medium, which cures overnight to form a hard seal, eliminating the need to seal the edges of the coverslips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty