Would you like to log in again?
CRITERION™ Tryptic Soy Broth #2, Hardy Diagnostics
Supplier: Hardy Diagnostics
CRITERION™
C9143 C9142 C9141 C9140
76461-860EA 114.85 USD InStock
76461-860 76461-862 76461-858 76461-864
CRITERION™ Tryptic Soy Broth #2, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Hardy Diagnostics CRITERION™ Tryptic Soy Broth #2 growth medium for cultivating fastidious and nonfastidious microorganisms. Tryptic Soy Broth #2 is formulated to be cold filterable, which means the media will go into solution at room temperature. Often used for media-fill challenges, non-sterile.
  • Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers - for efficient storage
  • Increased solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • Competitive pricing - call for a price quote for large volume purchases
  • Complete technical/instruction sheets - available in HUGO software
  • Unparalleled performance - tested for optimal growth
Container sizes include:
2 L mylar® bag – pre-measured for a 2 liter batch, no weighing needed
500 gm bottle - wide mouth, easy grip bottle, comes with a free scoop
2 kg and 10 kg bucket - with convenient screw lid
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty