Would you like to log in again?
Kleenguard™ A50 Breathable Splash and Particle Protection Coveralls, with Hood, Kimberly-Clark Professional
Supplier: Kimberly-Clark
KleenGuard®
51925 51929 51925 51928 51928 51927 51929 51926 51927 51926
76469-438CS 231.56 USD InStock
CA76469-438 76469-438 CA76469-436 CA76469-434 76469-434 76469-436 76469-442 CA76469-442 CA76469-440 76469-440
Kleenguard™ A50 Breathable Splash and Particle Protection Coveralls, with Hood, Kimberly-Clark Professional
Disposable Garments
KleenGuard™ A50 Coveralls are made of SMS fabric that’s been treated with a proprietary coating to offer both breathability and liquid splash protection in a single coverall.
  • KleenGuard™ A50 Coveralls are designed to protect against dry particles, aerosols, and chemical splash
  • Coveralls use Reflex® sizing for a more comfortable fit
  • Respirator fit hood
  • Two-way zipper offers easy access to work clothes
  • Thumb loops for better wrist protection
  • Elastic wrists and ankles
  • Individually packed
Breathable splash and particle protection coveralls with respirator fit hood. Reflex® sizing provides a more comfortable fit and ensures less rip outs. Passes EN1149-5 criteria for anti-static materials. Passes testing for Category III, Type 5, and Type 6 protection. Passes EN 1073-2 approved for radioactive dust protection. Thumb loops prevent sleeves from riding up and provide increase wrist protection.
Certifications: EN 1149-5 for antistatic materials. EN 1073-2 approved for radioactive dust protection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty