Would you like to log in again?
Oakton® Ion 6+ Meters, Cole-Parmer®
Supplier: Cole Parmer
Oakton®
35613-82 35613-80
76221-254EA 860.98 USD InStock
76221-254 76221-256
Oakton® Ion 6+ Meters, Cole-Parmer®
Electrochemical Meters pH and Multi-Parameter Meters
Direct readout of ion concentration at an economical price.
  • Compact and easy-to-use with preprogrammed ion calibration points
  • mV mode allows you to check electrode calibration and performance
  • ±1999 mV range is compatible with virtually all ISE electrodes
These rugged, economical meters are very easy to use. Features include all push-button calibration, hold function, auto-off, and splash-resistant keypad. Read concentration in ppm and mV as well as pH and temperature in °C.
Ordering information: Order ISE electrodes separately.
Delivery information: The included protective rubber boot also acts as a meter stand for benchtop use.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty