Would you like to log in again?
SterEM™ R2A Agar, Irradiated, Hardy Diagnostics
Supplier: Hardy Diagnostics
W530
76443-238PK 51.5 USD InStock
76443-238
SterEM™ R2A Agar, Irradiated, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Irradiated R2A Agar is recommended for enumeration of heterotrophic bacteria in water, especially potable water.
  • New moisture control packaging format prevents moisture buildup and ensures safe and sterile transfer
  • Optically clear polystyrene for distortion free viewing
  • Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
  • Sterile by irradiation validated for Sterility Assurance Level (SAL) of 10-5
  • Two nylon inner bags. One outer Mylar® bag that is heat sealed
  • Certificated of Analysis available at the Hardy Diagnostics website 24/7
Certifications: Manufactured in a licensed cGMP facility with the quality management system certified to ISO 13485.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty