Would you like to log in again?
Tryptic Soy Agar (TSA), pH 8.0, Hardy Diagnostics
Supplier: Hardy Diagnostics
G608
76443-242PK 33.5 USD InStock
76443-242
Tryptic Soy Agar (TSA), pH 8.0, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For use as a general growth medium for the cultivation and maintenance of microorganisms from industrial samples.
  • Optically clear polystyrene for distortion free viewing
  • Heavy-Duty - Reduces breakage during shipping
  • Stacking rings guard against slippage when moving stacked plates
  • Packaged in sleeves of 10 plates in a 'breathable' bag that prevents buildup of condensation and excess moisture
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty