Would you like to log in again?
Colitag™ Sample Containers, Hardy Diagnostics
Supplier: Hardy Diagnostics
Colitag™
9855
76443-240PK 107.01 USD InStock
76443-240
Colitag™ Sample Containers, Hardy Diagnostics
Sample Containers General Purpose Containers
Sterile coliform bottles with a 120 ml volume to meet EPA requirements.
100 ml mark molded into the polypropylene plastic, encircling the bottle to make it easy to see and feel the fill line.
The bottle contains has a tamper proof seal to ensure sterilization.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty