Would you like to log in again?
PLET Agar, for Bacillus anthracis, Hardy Diagnostics
Supplier: Hardy Diagnostics
G153
76443-248PK 62.5 USD InStock
76443-248
PLET Agar, for Bacillus anthracis, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of Bacillus anthracis from environmental samples, animal products, carcasses, and clinical samples from non-sterile sites.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty