Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mid-Turbinate Adult Flocked Swab With Stopper, C&A Scientific
Supplier: C&A Scientific
972014
76446-898EA 96.88 USD InStock
76446-898
Mid-Turbinate Adult Flocked Swab With Stopper, C&A Scientific
Swabs Swabs for Clinical Use
Hardy Diagnostics offers premium flocked swabs.
  • Highly absorbent tip
  • Soft smooth, non-abrasive
Our swabs ensure maximum collection and release of specimens, specifically for applications in relation to COVID-19. With integrated 80mm breakpoint on our flocked swab, it is easily paired with our Viral Transport Medium (Cat. no. 76334-640) for the preservation of viral specimens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty