Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 122 mm Probe

Supplier: VWR
VWR®
60900-3711
76472-450CS 575.53 USD InStock
76472-450
VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 122 mm Probe
Thermometers Digital Thermometers
Surface probe stem thermometer is ideal for cooking, laboratories and hot plates.

  • Stainless-steel 120 mm (4.7") probe keep hand away from hot surfaces
  • Will read temperatures in liquids, semi-solids and gases
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialized, CE marked

Certifications: Tested and calibrated in exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.

Delivery information: Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish).

Caution: These electronic stem thermometers are not suitable for solids such as frozen items.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty