Would you like to log in again?
Eisco® Stainless Steel Hanger for Slotted Weights
Supplier: EISCO SCIENTIFIC LLC
PHSSH050
470332-238EA 6.3 USD InStock
470332-238
Eisco® Stainless Steel Hanger for Slotted Weights
Educational Materials Physics Educational Materials Mechanics Learning Activities
Replacement hanger for your stainless steel slotted weight set.
  • For use with stainless steel slotted masses up to 50 g
  • Total capacity up to 500 g
This premium quality, durable stainless steel slotted masses hanger is for use with stainless steel slotted masses weighing up to 50 g. Hanger is 5.5" tall with top hook and a 1.25" Ø base for extra stability.
Ordering information: Replacement hanger only; slotted masses not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty