Would you like to log in again?
Tissue Pulverizer, 40355, Cole Parmer
Supplier: Cole Parmer
36903-05
MSPP-WU3690305EA 744.23 USD InStock
MSPP-WU3690305
Tissue Pulverizer, 40355, Cole Parmer
Homogenizers Tissue Grinders
Ideal for freeze-fracturing tough tissues like skin, cartilage, cornea, hair, nails and fresh bone.
  • 316 stainless steel construction designed for liquid nitrogen temperature
This tissue pulverizer reduces samples frozen in liquid nitrogen to a coarse powder. Use this powder as a preliminary step for cell disruption (homogenization) and metabolite, protein and nucleic acid extraction. Available in two sizes for tissues samples of 10 mg to 1 g.
Dispersion Type: Coarse
Sampling size: 0.1 to 1 g (tissue)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty